Learn More
YSI 3P LifeScience ELECTROLYTE FOR GALVANIC SENSO
Catalog No. 50465213
Encompass_Preferred
Electrolyte for galvanic oxygen sensors StirrOx G, CellOx 325, DurOx 325, TA 197 Oxi ConOx and MPP 350 Ox module, 1 bottle with 50 ml.
Catalog No. 50-465-213 Supplier YSI 3P LifeScience Supplier No. 205217
Electrolyte for galvanic oxygen sensors StirrOx G, CellOx 325, DurOx 325, TA 197 Oxi ConOx and MPP 350 Ox module, 1 bottle with 50 ml.