Learn More
Hach Company Electrolyte for Oxygen Sensors, 2 x 25 mL - Refill for dissolved-oxygen sensor assemblies
Catalog No. NC1964091 Shop All Hach Company Products
Encompass
Electrolyte for oxygen sensors supplied as two 25 mL bottles. This refill electrolyte is used to replenish compatible dissolved-oxygen sensor assemblies and helps maintain measurement accuracy and sensor performance. The product is classified as hazardous; consult the safety data sheet (SDS) and follow appropriate handling procedures.
  • Package contains two 25 mL bottles for easy refilling.
  • Helps maintain sensor accuracy and extend sensor life.
  • Simple application process to minimize maintenance downtime.
  • Small volume suitable for periodic sensor maintenance.
  • Classified as hazardous-use appropriate personal protective equipment.
Catalog No. NC1964091 Supplier Hach Company Supplier No. 2959GP
Electrolyte for oxygen sensors supplied as two 25 mL bottles. This refill electrolyte is used to replenish compatible dissolved-oxygen sensor assemblies and helps maintain measurement accuracy and sensor performance. The product is classified as hazardous; consult the safety data sheet (SDS) and follow appropriate handling procedures.
  • Package contains two 25 mL bottles for easy refilling.
  • Helps maintain sensor accuracy and extend sensor life.
  • Simple application process to minimize maintenance downtime.
  • Small volume suitable for periodic sensor maintenance.
  • Classified as hazardous-use appropriate personal protective equipment.