Learn More
MSC MCR Safety Checklite CL1 Safety Glasses, Polycarbonate, Clear Lens, 1 Pair
Catalog No. 5000684849
Encompass
These safety glasses are engineered for everyday eye protection, offering a blend of comfort, durability, and a precise orbital seal. With a lightweight construction of 22.9 g, they are suitable for both daily tasks and visitor use. The design features a smaller frame for a secure fit on smaller face sizes and high-strength polycarbonate lenses that provide reliable protection against environmental hazards.
  • Features durable polycarbonate lenses that filter 99.9% of harmful UV rays
  • Equipped with a scratch-resistant coating for enhanced clarity and longevity
  • Meets ANSI/ISEA Z87.1-2025, CAN/CSA Z94.3, CE EN166, and AS/NZS 1337.1 safety standards
  • Passes MIL-PRF-31013 ballistic standards for superior impact resistance
  • Lightweight design weighing less than 23 g for long-lasting comfort
  • Spatula-style temples and integrated nosebridge constructed from polycarbonate
Catalog No. 50-006-84849 Supplier MSC Supplier No. 52408119
These safety glasses are engineered for everyday eye protection, offering a blend of comfort, durability, and a precise orbital seal. With a lightweight construction of 22.9 g, they are suitable for both daily tasks and visitor use. The design features a smaller frame for a secure fit on smaller face sizes and high-strength polycarbonate lenses that provide reliable protection against environmental hazards.
  • Features durable polycarbonate lenses that filter 99.9% of harmful UV rays
  • Equipped with a scratch-resistant coating for enhanced clarity and longevity
  • Meets ANSI/ISEA Z87.1-2025, CAN/CSA Z94.3, CE EN166, and AS/NZS 1337.1 safety standards
  • Passes MIL-PRF-31013 ballistic standards for superior impact resistance
  • Lightweight design weighing less than 23 g for long-lasting comfort
  • Spatula-style temples and integrated nosebridge constructed from polycarbonate