Learn More
Hach Company ELECTROLYTE RE-FILL FOR A 11ML
Catalog No. NC1916637 Shop All Hach Company Products
Encompass
Electrolyte re-fill for AMTAX sc GSE electrode, 11ml. Electrolyte refill to be used with all ranges.
This item is not returnable. View return policy
Catalog No. NC1916637 Supplier Hach Company Supplier No. 2894536
Electrolyte re-fill for AMTAX sc GSE electrode, 11ml. Electrolyte refill to be used with all ranges.