Learn More
Hach Company Hach Amtax SC Electrode Set - 3 electrolyte solutions and 3 membrane caps (Range 1, 0.02-5.0 mg/L NH4-N)
Catalog No. NC3496260 Shop All Hach Company Products
Encompass
Electrode set for servicing ammonium electrodes in online analyzers; contains three electrolyte refill solutions and three membrane caps to restore electrode performance and response.
  • Contains three electrolyte solutions and three membrane caps.
  • Restores electrode response and accuracy after maintenance.
  • Suitable for ammonium measurement in the 0.02-5.0 mg/L NH4-N range.
  • Intended for electrode maintenance and replacement in online analyzers.
Catalog No. NC3496260 Supplier Hach Company Supplier No. 2955300
Electrode set for servicing ammonium electrodes in online analyzers; contains three electrolyte refill solutions and three membrane caps to restore electrode performance and response.
  • Contains three electrolyte solutions and three membrane caps.
  • Restores electrode response and accuracy after maintenance.
  • Suitable for ammonium measurement in the 0.02-5.0 mg/L NH4-N range.
  • Intended for electrode maintenance and replacement in online analyzers.