Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrolyte Solution, Nonaqueous, 2 M LiCl in Ethanol, MilliporeSigma™ Supelco™

Catalog No. 111019605 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9605 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9605 Supplier MilliporeSigma Supelco Supplier No. 56754250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Filling solution for reference electrodes used for titrations in nonaqueous solutions.

  • Concentration: 2 M LiCl in ethanol
  • Application: Filling solution for reference electrodes used for titrations in nonaqueous solutions
  • Quality: Nonaqueous
  • Molecular Weight: 42.39
  • Flash Point: 53.6 °F
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Chloride
CAS 7447-41-8
Physical Form Liquid
Quantity 250 mL
Linear Formula LiCl
MDL Number MFCD00011078
UN Number UN1170
Flash Point 12°C (53.6°F)
Synonym Lithium chloride solution
InChI Formula InChI=1S/ClH.Li/h1H;/q;+1/p-1
InChI Key KWGKDlikayfuFQ-UHFFFAOYSA-M
SMILES CCO.[Li+].[Cl-]
IUPAC Name Ethanol Lithium Chloride
Molecular Weight (g/mol) 42.39
PubChem CID 11235
Formula Weight 42.39
Concentration or Composition (by Analyte or Components) 2 M LiCl in ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.