Learn More
Verichem Laboratories ELECTROLYTE STANDARD KIT
Catalog No. NC9838976
Encompass_Preferred
Protein-based, liquid standards Intended for calibration verification or calibration Suitable for most direct ISE, diluted ISE and cesium flame Glycol and surfactant free for maximum electrode protection Revised formulation for lithium direct ISE
Catalog No. NC9838976 Supplier Verichem Laboratories Supplier No. 9200
Protein-based, liquid standards Intended for calibration verification or calibration Suitable for most direct ISE, diluted ISE and cesium flame Glycol and surfactant free for maximum electrode protection Revised formulation for lithium direct ISE