Learn More
GSC Go Science Crazy Electromagnet
Catalog No. S24647 Shop All GSC Products
Catalog No. S24647 Supplier GSC Go Science Crazy Supplier No. 5053
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Iron Clad Electromagnet
Our iron clad electromagnet demonstrates how electrical current can be used to produce a strong magnetic field. This precision machined unit has tremendous lifting power. The electromagnet consists of a core and yoke with 4 D-cell battery holders. The core of the unit has a cylindrical coil wound with enameled copper wire. The unit can also operate on a 6V/2A DC power supply.