Description
Height-adjustable swivel casters for horizontal alignment of unit on uneven floors.
Specifications
Specifications
| For Use With (Equipment) | KBF Series |
| Material | Steel |
| Type | Castor Frame |