Learn More
Grainger Intermatic Mechanical Control Panel, 100 A, 8-Breaker Spaces, NEMA 3R, 1 ea
Catalog No. 502795034
Encompass_Preferred
The Intermatic load center is an all-in-one electrical control center designed for pool and spa installations. It features two T104M mechanical timer mechanisms and provides eight breaker spaces, eliminating the need for additional service panels or junction boxes. The enclosure is rated NEMA 3R for indoor and outdoor use, offering a robust steel construction with a beige finish.
  • Features two T104M DPST time switch mechanisms
  • Provides eight breaker spaces for branch circuit protection
  • Rated 100 A at 120/240 V AC
  • NEMA 3R rainproof enclosure for versatile indoor and outdoor installation
  • Compatible with pool, spa, and fountain applications including pumps and lighting
  • Includes mounting brackets and hardware for ease of installation
  • Certified to cCSAus standards
Catalog No. 50-279-5034 Supplier Grainger Supplier No. 1A850
The Intermatic load center is an all-in-one electrical control center designed for pool and spa installations. It features two T104M mechanical timer mechanisms and provides eight breaker spaces, eliminating the need for additional service panels or junction boxes. The enclosure is rated NEMA 3R for indoor and outdoor use, offering a robust steel construction with a beige finish.
  • Features two T104M DPST time switch mechanisms
  • Provides eight breaker spaces for branch circuit protection
  • Rated 100 A at 120/240 V AC
  • NEMA 3R rainproof enclosure for versatile indoor and outdoor installation
  • Compatible with pool, spa, and fountain applications including pumps and lighting
  • Includes mounting brackets and hardware for ease of installation
  • Certified to cCSAus standards