Learn More
MSC Sprayon Contact Cleaner, Flammable, Safe for Most Plastics, 16 oz
Catalog No. 502870143
Encompass
This product is a flammable contact cleaner designed for electrical and electronic applications. It is supplied in an aerosol can and formulated to be safe for use on most plastics, making it suitable for a wide range of devices.
  • Formulated to be safe for most plastics
  • Liquid form for effective cleaning and degreasing
  • Certified NSF K2 Registered for specific applications
  • Suitable for meters, hydraulic systems, TVs, timing devices, relays & switches, printed circuits, and other electronic equipment
  • Maximum temperature resistance of 70 F
  • Supplied in a 16 oz aerosol can with 11.00 oz net fill
Catalog No. 50-287-0143 Supplier MSC Supplier No. 84253871
This product is a flammable contact cleaner designed for electrical and electronic applications. It is supplied in an aerosol can and formulated to be safe for use on most plastics, making it suitable for a wide range of devices.
  • Formulated to be safe for most plastics
  • Liquid form for effective cleaning and degreasing
  • Certified NSF K2 Registered for specific applications
  • Suitable for meters, hydraulic systems, TVs, timing devices, relays & switches, printed circuits, and other electronic equipment
  • Maximum temperature resistance of 70 F
  • Supplied in a 16 oz aerosol can with 11.00 oz net fill