Learn More
MSC Sprayon Electrical Grade Cleaner, 39500V Dielectric Strength, Aerosol, 19 oz
Catalog No. 502870142
Encompass
Sprayon Electrical Grade Cleaner is a non-flammable solvent formulated for cleaning electrical and electronic components. This cleaner comes in a 19 oz aerosol can, offering a high dielectric strength of 39,500V for safe application. It is not safe for use on most plastics and is not food grade.
  • High 39,500V dielectric strength
  • Non-flammable formula
  • Supplied in a convenient aerosol can
  • Ideal for machinery, switches, motors, and printed circuits
  • SNAP Approved and NSF K2 Registered
  • RoHS Compliant
Catalog No. 50-287-0142 Supplier MSC Supplier No. 84253335
Sprayon Electrical Grade Cleaner is a non-flammable solvent formulated for cleaning electrical and electronic components. This cleaner comes in a 19 oz aerosol can, offering a high dielectric strength of 39,500V for safe application. It is not safe for use on most plastics and is not food grade.
  • High 39,500V dielectric strength
  • Non-flammable formula
  • Supplied in a convenient aerosol can
  • Ideal for machinery, switches, motors, and printed circuits
  • SNAP Approved and NSF K2 Registered
  • RoHS Compliant