Learn More
Electron Microscopy Sciences Cryo Markers
SDP
Catalog No. NC1562022
Encompass_Preferred
For permanently marking cloth, cardboard, and plastic. It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).
Catalog No. NC1562022 Supplier Electron Microscopy Sciences Supplier No. 62050
For permanently marking cloth, cardboard, and plastic. It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).