Learn More
Thermo Scientific Electron Multiplier
Catalog No. 501346563 Shop All Thermo Scientific Products
Encompass
Electron multiplier™ is vacuum tube used to amplify electron beam by utilizing secondary emission, where incident electrons strike dynode surface and cause emission of multiple secondary electrons; these secondary electrons are then accelerated and focused onto subsequent dynodes, resulting in exponential increase in electron current, providing high gain while maintaining low noise levels, making it useful for detecting and measuring low-level currents in mass spectrometers.
Catalog No. 50-134-6563 Supplier Thermo Scientific Supplier No. 1R7602214633
Electron multiplier™ is vacuum tube used to amplify electron beam by utilizing secondary emission, where incident electrons strike dynode surface and cause emission of multiple secondary electrons; these secondary electrons are then accelerated and focused onto subsequent dynodes, resulting in exponential increase in electron current, providing high gain while maintaining low noise levels, making it useful for detecting and measuring low-level currents in mass spectrometers.