Learn More
Cell Signaling Technology Electron Transport Chain Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502726373
Encompass
The Electron Transport Chain (Complex I, III, IV) Antibody Sampler Kit provides an economical solution for the detection of select components involved in mitochondrial electron transport chain complexes. The kit includes a curated selection of antibodies targeting key proteins in Complexes I, III, and IV, facilitating comprehensive analysis of mitochondrial respiratory chain function.
  • Economical format includes sufficient antibody volume to perform two western blot experiments with each primary antibody
  • Includes reagents for detecting proteins essential for energy metabolism and apoptotic signaling
  • Validated for multiple applications, including western blotting, immunofluorescence, and immunoprecipitation
  • Supplied with an HRP-linked secondary antibody for efficient signal detection
Catalog No. 50-272-6373 Supplier Cell Signaling Technology Supplier No. 42642T
The Electron Transport Chain (Complex I, III, IV) Antibody Sampler Kit provides an economical solution for the detection of select components involved in mitochondrial electron transport chain complexes. The kit includes a curated selection of antibodies targeting key proteins in Complexes I, III, and IV, facilitating comprehensive analysis of mitochondrial respiratory chain function.
  • Economical format includes sufficient antibody volume to perform two western blot experiments with each primary antibody
  • Includes reagents for detecting proteins essential for energy metabolism and apoptotic signaling
  • Validated for multiple applications, including western blotting, immunofluorescence, and immunoprecipitation
  • Supplied with an HRP-linked secondary antibody for efficient signal detection