Learn More
Troemner™ Electronic Balance ASTM Class 2 Weight
Catalog No. 02218366 Shop All Troemner Products
Change view
Click to view available options
Electronic Balance Weight; Weight: 5 kg; Material: Stainless Steel (303)
Catalog No.
02-218-366 Electronic Balance Weight; Weight: 5 kg; Material: Stainless Steel (303)
1 options
Catalog No. 02-218-366 Supplier Troemner™ Supplier No. 80861226
Add to Cart
Add to Cart
Troemner Individual Calibration Weights Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances.
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance No Certificate
Material Stainless Steel (303)
Case Material Polycarbonate
Weight (Metric) 5 kg
Shape Cylindrical
Includes Protective Case