Learn More
Troemner™ Electronic Balance ASTM Class 2 Weight with ISO/IEC 17025 Accredited Certificate
Catalog No. 02218370 Shop All Troemner Products
Change view
Click to view available options
Weight (Metric):
2 kg
5 kg
Shape:
Cylindrical
Grip Handle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric) Shape
02-218-370 2 kg Cylindrical
02-218-368 5 kg Cylindrical
02-218-365 5 kg Grip Handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-218-370 Supplier Troemner™ Supplier No. 80861240
Add to Cart
Add to Cart
Troemner Individual Calibration Weights Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances.
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance ISO/IEC 17025 Accredited Certificate
Material Stainless Steel (303)
Case Material Polycarbonate
Weight (Metric) 2 kg
Shape Cylindrical
Includes Protective Case