Learn More
Troemner™ Electronic Balance ASTM Class 2 Weight with Traceable Certificate
Catalog No. 02218362 Shop All Troemner Products
Change view
Click to view available options
Electronic Balance Weight; Weight: 10 kg
Catalog No.
02-218-362 Electronic Balance Weight; Weight: 10 kg
1 options
Catalog No. 02-218-362 Supplier Troemner™ Supplier No. 80861215
Add to Cart
Add to Cart
Troemner Individual Calibration Weights Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances.
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance Traceable Certificate
Material Stainless Steel (303)
Weight (Metric) 10 kg
Shape Grip Handle