Learn More
Troemner™ Electronic Balance ASTM Class 3 Weight with ISO/IEC 17025 Accredited Certificate
Catalog No. 02218403 Shop All Troemner Products
Change view
Click to view available options
Weight (Metric):
100 g
200 g
300 g
500 g
1000 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
02-218-403 300 g
02-218-407 100 g
02-218-405 200 g
02-218-401 500 g
02-218-399 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-218-403 Supplier Troemner™ Supplier No. 80861414
Only null left
Add to Cart
Add to Cart
Troemner Individual Calibration Weights Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances.
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance ISO/IEC 17025 Accredited Certificate
Material Stainless Steel (316)
Case Material Polycarbonate
Weight (Metric) 300 g
Shape Cylindrical
Includes Protective Case