Learn More
Grainger Electronic Digital Outside Mic
Catalog No. 502766266
Encompass_Preferred
The Mitutoyo IP65 Digital Outside Micrometer is a precision measuring instrument for outside dimensions. It has a measuring range of 0-25mm/0-1in with a resolution of 0.001mm/0.00005in. The micrometer has an ABSOLUTE linear encoder that provides resistance to shocks and vibrations. It is designed with IP65 protection making it dust/water resistant. The micrometer has a ratchet stop for consistent measuring force and preset to 30-50N of measuring force. It runs on SR44 silver oxide battery. The micrometer comes with inspection certificate.
Catalog No. 50-276-6266 Supplier Grainger Supplier No. 16X240
The Mitutoyo IP65 Digital Outside Micrometer is a precision measuring instrument for outside dimensions. It has a measuring range of 0-25mm/0-1in with a resolution of 0.001mm/0.00005in. The micrometer has an ABSOLUTE linear encoder that provides resistance to shocks and vibrations. It is designed with IP65 protection making it dust/water resistant. The micrometer has a ratchet stop for consistent measuring force and preset to 30-50N of measuring force. It runs on SR44 silver oxide battery. The micrometer comes with inspection certificate.