Learn More
MSC Insize USA LLC Electronic Edge Finder, 0.4 in. Head, 3/4 in. Shank, 1 ea.
Catalog No. 5000516751
Encompass
This electronic edge finder is designed for precision machine setup and location gaging on flat surfaces. It features a ball contact head to ensure accurate alignment during measurement tasks.
  • Features a 0.4 in. head diameter and a 3/4 in. shank diameter
  • Provides visual edge alert mechanism for clear identification
  • Constructed with a hardened shank and contact ball for durability
  • Operates using a 12V 23AE battery
  • Accuracy of 0.0002 in.
  • Not suitable for rotary use
Catalog No. 50-005-16751 Supplier MSC Supplier No. 28730588
This electronic edge finder is designed for precision machine setup and location gaging on flat surfaces. It features a ball contact head to ensure accurate alignment during measurement tasks.
  • Features a 0.4 in. head diameter and a 3/4 in. shank diameter
  • Provides visual edge alert mechanism for clear identification
  • Constructed with a hardened shank and contact ball for durability
  • Operates using a 12V 23AE battery
  • Accuracy of 0.0002 in.
  • Not suitable for rotary use