Learn More
Labsciences ELECTRONIC PIPETTE FILLER.
Catalog No. 50821779
Encompass_Preferred
Electronic pipette filler. Pipette filler has membrane filter and LED's to indicate charging status. Red collet accepts standard pipettes to 50ml. Rechargable NiCd batteries for 8 hours of operation. Boxed.
Catalog No. 50-821-779 Supplier Labsciences Supplier No. 110CREF
Electronic pipette filler. Pipette filler has membrane filter and LED's to indicate charging status. Red collet accepts standard pipettes to 50ml. Rechargable NiCd batteries for 8 hours of operation. Boxed.