Learn More
Labsciences ELECTRONIC PIPETTE FILLER.
Catalog No. 50821778
Encompass_Preferred
Electronic pipette filler. Pipette filler has membrane filter and LED's to indicate charging status. Yellow collet accepts standard pipettes to 50ml. Rechargable NiCd batteries for 8 hours of operation. Boxed.
Catalog No. 50-821-778 Supplier Labsciences Supplier No. 110CYEF
Add to Cart
Add to Cart
Electronic pipette filler. Pipette filler has membrane filter and LED's to indicate charging status. Yellow collet accepts standard pipettes to 50ml. Rechargable NiCd batteries for 8 hours of operation. Boxed.