Learn More
Labsciences ELECTRONIC PIPETTES.
Supplier:  Labsciences 10EP
Encompass_Preferred
Electronic pipette. Volume Range 0.2 to 10uL. Multi-mode operation, variable working speeds, simple volume selection. Large liquid crystal display, single fire button and integral tip ejector. Rechargable. Boxed.
Catalog No. 50-821-772