Learn More
Fisherbrand™ Electronic Stand
Catalog No. FB14955241 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
FB14955241 Acrylic
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. FB14955241
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Accessory for pipettes
  • Built-in battery system that provides consistent power for operation
  • Automatic charging capability
Specifications
Autoclavable Not Autoclavable
Color Clear/White
Holds Electronic Pipette
Material Acrylic
Positions 1
Shape Horizontal
Stackable No
Sterility Non-sterile
Product Type Pipette Holder and Charger
Shelves None