Learn More
Grainger Electronic Timer 7 Days (2) SPST-NO
Catalog No. 502750880
Encompass_Preferred
The OMRON Electronic Timer is a 7-day programmable timer that can be used to control lights, appliances, pumps, and more. It has up to 140 on/off settings per week and a manual override switch. The timer plugs into a standard wall outlet and has a grounded 3-prong outlet on the bottom to plug lights or appliances into. It has an easy-to-read LCD display that shows the programs, time, and status. The timer is ETL certified and rated for 15A/1875W. It measures 5.1 x 2.8 x 1.7 inches. This versatile 7-day programmable timer by OMRON can automate the operation of lights and appliances on customized daily and weekly schedules.
Catalog No. 50-275-0880 Supplier Grainger Supplier No. 14H249
The OMRON Electronic Timer is a 7-day programmable timer that can be used to control lights, appliances, pumps, and more. It has up to 140 on/off settings per week and a manual override switch. The timer plugs into a standard wall outlet and has a grounded 3-prong outlet on the bottom to plug lights or appliances into. It has an easy-to-read LCD display that shows the programs, time, and status. The timer is ETL certified and rated for 15A/1875W. It measures 5.1 x 2.8 x 1.7 inches. This versatile 7-day programmable timer by OMRON can automate the operation of lights and appliances on customized daily and weekly schedules.