Learn More
Stoelting™ Electronic Vaginal-Estrous Cycle-Monitor
SDP
Catalog No. 10001046 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Rat
Catalog No. Animal
10-001-046 Rat
1 options
Catalog No. 10-001-046 Supplier Stoelting™ Supplier No. 59160
Add to Cart
Add to Cart
Request a Quote
Electronic Vaginal-Estrous Cycle-Monitor is designed to obtain precise information to determine the optimum day for mating during the rat estrous cycle.
  • Electrical impedance of the epithelial cell layer of vaginal mucosa is measured at the frequency of 1 kHz by insertion of the probe into the vagina.
  • In the proestrus stage significantly higher impedance is produced compared to that in the other stages of the estrous cycle.
  • 3 kohm of impedance can be considered a standard for indicating proestrus stage.
  • Measuring range is 0 to 19.9 kohm
  • Ideal alternative to the conventional method (vaginal smear method) that can be a troublesome procedure requiring skill, time and labor
Specifications
For Use With (Application) Neuroscience
Animal Rat