Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Electrophoresis Buffers
SDPSCI_ED

Catalog No. S94619 Shop All EDVOTEK Products
Change view
Click to view available options
Chemical Name or Material:
10X TBE (Tris-borate-EDTA)
50x TAE (Tris-Acetate EDTA)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
S94619 10X TBE (Tris-borate-EDTA)
S11607 50x TAE (Tris-Acetate EDTA)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S94619 Supplier Edvotek™ Supplier No. 6071
Add to Cart
Add to Cart

Buffers for use in the science classroom.

  • This powdered blend Tris-Borate EDTA buffer (TBE), has been optimized for agarose gel electrophoresis of nucleic acids
  • Dissolve powder in 500 ml of distilled or deionized water to prepare a 10x concentrate which can be diluted 10-fold to make a 1x working electrophoresis buffer

Specifications

Chemical Name or Material 10X TBE (Tris-borate-EDTA)
CAS 77-86-1, 60-00-4, 6850-28-8
Concentration 50X
For Use With (Application) Electrophoresis
For Use With (Equipment) Instruments and Electrodes
pH 7.8 to 8.0
Physical Form Aqueous Solution
Quantity For 5 L
Solution Type Buffer
Percent Purity 95 to 99%
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.