Learn More
Electron Microscopy Sciences EMS MyGel™ Mini Electrophoresis Starter Kit, 230 V
SDP
Catalog No. 501927845
Encompass_Preferred
The EMS MyGel Mini Electrophoresis System is used for quick and easy gel casting without clamps or spacers. Place the tray in the casting stand and pour. The trays feature a dark contrast strip to help visualize wells while loading and lines that fluoresce under UV and blue light to track progress of the run. System includes a gel tank, power supply and two casting sets. Ideal for personal use, small laboratories or the classroom.
The safety lid is vented to help dissipate heat generated during electrophoresis, and features a viewing area above the gel. Current will only run through the system when the lid is properly in place on top of the gel box.
Trays and casting stands for both 10.5 x 6 cm and 5 x 6 cm gels are included with the MyGel™ Mini system, as are double-sided combs. The smaller tooth combs are compatible with multichannel pipettes. With Mini Digital Balance 100g and 100g of Agarose LE.
Catalog No. 50-192-7845 Supplier Electron Microscopy Sciences Supplier No. 8705604
The EMS MyGel Mini Electrophoresis System is used for quick and easy gel casting without clamps or spacers. Place the tray in the casting stand and pour. The trays feature a dark contrast strip to help visualize wells while loading and lines that fluoresce under UV and blue light to track progress of the run. System includes a gel tank, power supply and two casting sets. Ideal for personal use, small laboratories or the classroom.
The safety lid is vented to help dissipate heat generated during electrophoresis, and features a viewing area above the gel. Current will only run through the system when the lid is properly in place on top of the gel box.
Trays and casting stands for both 10.5 x 6 cm and 5 x 6 cm gels are included with the MyGel™ Mini system, as are double-sided combs. The smaller tooth combs are compatible with multichannel pipettes. With Mini Digital Balance 100g and 100g of Agarose LE.