Learn More
Biotium Electrophoresis Products Equipment DNA Gel Stains and Accessories Gel Imaging 1/EA
Catalog No. 5000501589
Encompass_Preferred
Glo-Plate White Photoactivation Device is a LED light box designed for developing the staining of gels stained with the visible blue DNA gel stain DNAzure
Catalog No. 50-005-01589 Supplier Biotium Supplier No. E90008
Glo-Plate White Photoactivation Device is a LED light box designed for developing the staining of gels stained with the visible blue DNA gel stain DNAzure