Learn More
Embi Tec RunOne Electrophoresis System, without Timer
SDP
Catalog No. NC9680743
Encompass
110 V RunOne Electrophoresis System 8/99,includes the RunOne Electrophoresis Cell and Power supply for 100/110V operation, the RunOne Casting System (6 compartment horizontal casting stand with lid, 6 agarose gel trays,2 reversible combs.)
Catalog No. NC9680743 Supplier Embi Tec Supplier No. EP2000
110 V RunOne Electrophoresis System 8/99,includes the RunOne Electrophoresis Cell and Power supply for 100/110V operation, the RunOne Casting System (6 compartment horizontal casting stand with lid, 6 agarose gel trays,2 reversible combs.)