Learn More
MSE Supplies LLC MSE PRO 700ml Western Blot Electrophoresis Tank
SDP
Supplier:  MSE Supplies LLC BB2369
Encompass
MSE PRO™ 700ml Western Blot Electrophoresis TankThe MSE PRO Western Blot Transfer Electrophoresis Tank is a high-efficiency system designed for the rapid and precise transfer of small protein gels. Engineered with advanced electrode positioning, it generates a strong electric field to ensure effective protein transfer. Its optional built-in ice box or blue crystal ice box effectively dissipates heat during electrophoresis, preventing gel degradation. With a safety-focused design, including automatic power-off and flexible wiring, this system offers reliability and ease of use for laboratories conducting protein analysis and research. Use with (BB2367, BB2368) and recommended power supply (BB2359, BB2361, BB2357).Optimized Electrode Design: Close electrode spacing (4 cm) enhances the electric field for efficient protein transfer.Advanced Cooling System: Optional ice box effectively absorbs heat, preserving sample integrity.Enhanced
Catalog No. NC3739822