Learn More
USA Scientific Inc Electroporation cuvette, .4 cm
Catalog No. 502745308
Encompass
Electroporation cuvette, 0.4 cm gap width, red cap, individually wrapped, str, 50/bag
Catalog No. 50-274-5308 Supplier USA Scientific Inc Supplier No. 91046050
Electroporation cuvette, 0.4 cm gap width, red cap, individually wrapped, str, 50/bag