Learn More
Alkali Scientific Electroporation Cuvettes Polyc
SDP
Catalog No. 503027712
Encompass_Preferred
Electroporation Cuvettes are used to transfer DNA RNA or proteins into multiple cell types including bacteria yeast mammalian and plant cell samples They are made from high quality durable polycarbonate material and are compatible with most electroporation devices Precision built to give exacting tolerances our Electroporation Cuvettes ensure reproducible results Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies Precision built to give exacting tolerances ensuring reproducible results Polished aluminum plates reduce static and arcing for optimum performance Individually wrapped and gamma irradiated providing guaranteed sterility Compatible with most Electroporation devices Individually color coded cuvette caps identify gap widths of 1mm 2mm and 4mm Each bag contains 50 individually wrapped sterile cuvettes What are the minimal and maximal operating volumes 1 mm gap cuvettes 20 to 100 L2 mm gap cuvettes 40 to 4
Catalog No. 50-302-7712 Supplier Alkali Scientific Supplier No. RP401001
Electroporation Cuvettes are used to transfer DNA RNA or proteins into multiple cell types including bacteria yeast mammalian and plant cell samples They are made from high quality durable polycarbonate material and are compatible with most electroporation devices Precision built to give exacting tolerances our Electroporation Cuvettes ensure reproducible results Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies Precision built to give exacting tolerances ensuring reproducible results Polished aluminum plates reduce static and arcing for optimum performance Individually wrapped and gamma irradiated providing guaranteed sterility Compatible with most Electroporation devices Individually color coded cuvette caps identify gap widths of 1mm 2mm and 4mm Each bag contains 50 individually wrapped sterile cuvettes What are the minimal and maximal operating volumes 1 mm gap cuvettes 20 to 100 L2 mm gap cuvettes 40 to 4