Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Electrostatic Kit
GREENER_CHOICESCI_ED

Catalog No. S160137 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160137 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160137 Supplier Supertek™ Supplier No. PH45053SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use this handy Supertek™ Electrostatic Kit for demonstrating the generation of electrostatic charges with friction rods.

  • Electrostatic kit with synthetic fur
  • Instructions included

Specifications

Grade 9 to Undergraduate
Format Kit
Includes 1 hollow glass rod, 1 solid glass rod, 1 hard rubber rod, 1 silk friction pad, 1 wool friction pad, 1 flannel friction pad, 1 animal fur( synthetic) friction pad, 3 balloons, 10 ft. of string, and instructions manual
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.