Learn More
World Precision Instrument Economy Electrosurgical Unit
SDP
Catalog No. 50822633
Encompass
Cut and coagulate with a press of the foot switch
Electrosurgery utilizes alternating current at radio frequencies to cut and coagulate. Using this method, the current enters the subject's body and the subject becomes part of the electrical circuit. This requires the use of a return, or ground, plate. This economically priced electrosurgical unit has 10 levels of output intensity, three operational modes (cut, coagulate, and cut/coagulate) and various choices of electrodes. The unit comes complete and ready-to-use with a handpiece, ground plate, foot switch and one of each electrodes. All accessories can also be ordered separately.
Catalog No. 50-822-633 Supplier World Precision Instrument Supplier No. 501274
Cut and coagulate with a press of the foot switch
Electrosurgery utilizes alternating current at radio frequencies to cut and coagulate. Using this method, the current enters the subject's body and the subject becomes part of the electrical circuit. This requires the use of a return, or ground, plate. This economically priced electrosurgical unit has 10 levels of output intensity, three operational modes (cut, coagulate, and cut/coagulate) and various choices of electrodes. The unit comes complete and ready-to-use with a handpiece, ground plate, foot switch and one of each electrodes. All accessories can also be ordered separately.