Learn More
Med Vet International Electrosurgical Unit Electrode Heavy Wire, J0470DT4
Catalog No. 5000583848
Encompass_Preferred
Autoclavable. 1/16" Use only with J-470 unitNOTE: DO NOT store in liquids..
Catalog No. 50-005-83848 Supplier Med Vet International Supplier No. JOR470DT4
Autoclavable. 1/16" Use only with J-470 unitNOTE: DO NOT store in liquids..