Learn More
Perkin Elmer US LLC Elegra Argon Humidifier
Catalog No. 502104159
Encompass_Preferred
This assembly includes: Elegra Humidifier Gas Connector Humidifier Gas Line, 1/4-28 Ratchet EzyLok Assembly EzyLok Kit Features and Benefits: Compact design. No heating or electric power required. Non-pressurized water reservoir. An easy-to-use bypass switch allows you to take the Elegra off-line without disconnecting argon lines. Highly efficient membrane humidification technology. Improved signal stability for samples with high TDS. Simple to use and maintain. Facilitates long, uninterrupted run times. Inert metal-free construction eliminates possibility of contamination. Rugged and durable polymer casing. Maximum and minimum fill marks ensure that you are always operating under optimum conditions.
Catalog No. 50-210-4159 Supplier Perkin Elmer US LLC Supplier No. N0781598
This assembly includes: Elegra Humidifier Gas Connector Humidifier Gas Line, 1/4-28 Ratchet EzyLok Assembly EzyLok Kit Features and Benefits: Compact design. No heating or electric power required. Non-pressurized water reservoir. An easy-to-use bypass switch allows you to take the Elegra off-line without disconnecting argon lines. Highly efficient membrane humidification technology. Improved signal stability for samples with high TDS. Simple to use and maintain. Facilitates long, uninterrupted run times. Inert metal-free construction eliminates possibility of contamination. Rugged and durable polymer casing. Maximum and minimum fill marks ensure that you are always operating under optimum conditions.