Learn More
Glass Expansion Elegra Membrane
SDP
Catalog No. NC2333649
Encompass_Preferred
Replacement Elegra membrane. Compatible with all Glass Expansion Single and Dual-Channel Elegra Argon Humidifiers.If the Elegra membrane gets damaged, appears discolored or is suspected of being dirty/clogged, it should be replaced. Frequently inspecting the water level, and changing the water often, can eliminate algal growth and unwanted accumulation of debris. Do not leave water in Elegra for extended periods of time if not being used. To properly maintain components, if the Elegra Argon Humidifier is not used for long periods of time it is best to empty the water and dry components for storage.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2333649 Supplier Glass Expansion Supplier No. 708031286
Replacement Elegra membrane. Compatible with all Glass Expansion Single and Dual-Channel Elegra Argon Humidifiers.If the Elegra membrane gets damaged, appears discolored or is suspected of being dirty/clogged, it should be replaced. Frequently inspecting the water level, and changing the water often, can eliminate algal growth and unwanted accumulation of debris. Do not leave water in Elegra for extended periods of time if not being used. To properly maintain components, if the Elegra Argon Humidifier is not used for long periods of time it is best to empty the water and dry components for storage.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001