Learn More
Grainger Element Only 5-13/16 L
Catalog No. 502750314
Encompass_Preferred
The Baldwin Filters B1440 oil filter has a 5-13/16 inch overall length and 3/4 inch NPT thread size. It has a 90 psi anti-drainback valve, ideal for conventional oil. This full-flow cartridge oil filter fits many domestic vehicles and light trucks, including certain models of Buick, Cadillac, Chevrolet, Chrysler, Dodge, Ford, GMC, Jeep, Lincoln, Mercury, and Pontiac. The filtration media is cellulose and polyester, with a steel base plate and housing.
Catalog No. 50-275-0314 Supplier Grainger Supplier No. 14R259
The Baldwin Filters B1440 oil filter has a 5-13/16 inch overall length and 3/4 inch NPT thread size. It has a 90 psi anti-drainback valve, ideal for conventional oil. This full-flow cartridge oil filter fits many domestic vehicles and light trucks, including certain models of Buick, Cadillac, Chevrolet, Chrysler, Dodge, Ford, GMC, Jeep, Lincoln, Mercury, and Pontiac. The filtration media is cellulose and polyester, with a steel base plate and housing.