Learn More
Electron Microscopy Sciences Element Standard Storage Box, 20 Discs
SDP
Catalog No. 5028432
Encompass_Preferred
A sample storage box that holds either 10 or 20 of 1/4" diameter discs. It is designed to store the element discs securely; therefore, avoiding any contact with some of the dangerous elements. Holds 20.
Catalog No. 50-284-32 Supplier Electron Microscopy Sciences Supplier No. 8006320
A sample storage box that holds either 10 or 20 of 1/4" diameter discs. It is designed to store the element discs securely; therefore, avoiding any contact with some of the dangerous elements. Holds 20.