Learn More
Diversified Spaces™ Elementary Four-Student Table
SCI_ED
Catalog No. S42472 Shop All Diversified Spaces Products
Change view
Click to view available options
Material:
Maple
Maple, Laminate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S42472 Maple
S42474 Maple, Laminate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S42472 Supplier Diversified Spaces™ Supplier No. WX4M26M
Only null left
Add to Cart
Add to Cart
Designed to provide structurally sound and completely functional work center.
  • Constructed from solid maple and maple veneer
  • 2.063 x 11.43cm (0.8125 x 4.5 in.) solid maple aprons with 13-gauge steel corner braces
  • 5.715cm (2.25 in.) maple legs have adjustable floor glides and rubber leg boots
  • Finished with durable chemical resistant non-emitting UV finish
  • SEFA compliant and MAS Certified Green
Specifications
Product Type Lab Table
No. of Shelves None
Color Maple
Autoclavable Not Autoclavable
No. of Doors None
No. of Drawers None
With Casters No
Footrest No
With Tilt No
Top Style Maple
Warranty Limited Lifetime
Dimensions (D x W x H) 121.92 x 121.92 x 66.04 cm (48 x 48 x 26 in.)
Material Maple