Learn More
Supertek™ Elementary Microscope and Components Kit
GREENER_CHOICESCI_ED
Catalog No. S00680 Shop All Supertek Scientific Products
Change view
Click to view available options
Head Style:
Monocular
Catalog No. Head Style
S00680 Monocular
1 options
Catalog No. S00680 Supplier Supertek™ Supplier No. B10516S3
Add to Cart
Add to Cart
This lighted microscope is perfect for beginners. It features a 10x eyepiece, 10x, 20x, and 30x objectives, rack and pinion geared focusing.
  • Lighted microscope perfect for beginners.
  • 10x eyepiece
  • 10x, 20x, and 30x objectives
  • Rack & pinion geared focusing
  • Glass optics
  • Stage clips
  • Reflective mirror or battery operated illumination
Specifications
Eyepiece 10X
Head Style Monocular
Lighting Type Battery
Magnification Power 10X, 20X, 30X
Model B10516-S3
Objective Types 10x, 20x, 30x
Product Type Beginner Microscope