Learn More
Fisher Science Education™ Elementary Series Stereo Microscope
SCI_ED
Catalog No. S72589 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
2X
Catalog No. Magnification Power
S72589 2X
1 options
Catalog No. S72589
Stereoscopic Microscope for Macro-Size Specimens is ideal for elementary school classrooms
Optical System
  • 10X WF eyepiece
  • Fixed magnification
  • 2X objective
Viewing Head
  • Vertical Binocular viewing tubes
  • Interpupillary adjustment
  • Diopter Adjustment
Focusing
  • Coarse focus adjustment
  • 40mm height adjustment range on focusing mechanism
  • 70mm working distance
Stand
  • Low profile plain stage with clips
  • Pole stand
  • Compact design
Warranty and Services
Limited Lifetime Warranty
Specifications
Magnification Power 2X
Objective Types Fixed
Grade Elementary level
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Product Type Stereoscopic Microscope
Head Style Binocular
Eyepiece WF 10X
For the specifics on a particular model, see the specification table located at the bottom of the page of that individual model