Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elementary Snap Circuits Kit
SDPSCI_ED

Catalog No. S68214
Encompass
Encompass_Preferred
Change view
Click to view available options
Includes:
Storage case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S68214 Storage case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S68214 Supplier Elenco Electronics Inc Supplier No. SC100R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Storage case

Requires

Two AA batteries (S43921)

Great introduction to the world of electricity and electronics.

  • More than 30 parts easily snap together
  • Build over 100 experiments without any additional tools
  • Full-color manual includes information about resistors, capacitors, transistors, motors, batteries, series/parallel circuits and more

Specifications

Product Type Snap Circuits™ Student Guide
For Use With (Application) Great introduction to the world of electricity and electronics
Includes Storage case
Weight (English) Shipping 2.8 lb.
Weight (Metric) Shipping 1.27 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.