Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Elements, Mixtures and Compounds Lab Investigation
SCI_ED

Catalog No. S63468
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S63468 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S63468 Supplier Neo/Sci Corporation Supplier No. 201673

missing translation for 'validMainframeMsg'

Provides a basic understanding of the primary concepts of chemistry.

  • Investigate how matter is classified
  • Identify various types of mixtures
  • Compare the properties of solutions and mixtures
  • Identify elements and compounds
  • Study how chemical symbols, formulas and equations are used
  • Class size: 40 students

Specifications

For Use With (Application) Provides a basic understanding of the primary concepts of chemistry
Includes Individual software
Class Size 40 Students
Product Type Elements, Mixtures and Compounds Lab Investigation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.