Learn More
Apexbio Technology LLC β-Elemonic acid 28282-25-9 10mg
SDP
Catalog No. 5000094941
Encompass_Preferred
-Elemonic acid (CAS 28282-25-9) is a triterpenoid compound isolated from frankincense Experimental studies indicate that -elemonic acid modulates cellular apoptosis and redox processes in part by inducing reactive oxygen species (ROS) generation and upregulating cyclooxygenase-2 (COX-2) expression It also exhibits inhibitory activity against prolyl endopeptidase Owing to its observed anti-cancer and anti-inflammatory properties -elemonic acid serves as a valuable molecular probe in the investigation of disease models involving oxidative stress inflammation and apoptosis regulation
Catalog No. 50-000-94941 Supplier Apexbio Technology LLC Supplier No. A463310
Add to Cart
Add to Cart
-Elemonic acid (CAS 28282-25-9) is a triterpenoid compound isolated from frankincense Experimental studies indicate that -elemonic acid modulates cellular apoptosis and redox processes in part by inducing reactive oxygen species (ROS) generation and upregulating cyclooxygenase-2 (COX-2) expression It also exhibits inhibitory activity against prolyl endopeptidase Owing to its observed anti-cancer and anti-inflammatory properties -elemonic acid serves as a valuable molecular probe in the investigation of disease models involving oxidative stress inflammation and apoptosis regulation