Learn More
Medchemexpress LLC HY-109055 10mg Medchemexpress, Elenbecestat CAS:1388651-30-6 Purity:>98%
SDP
Catalog No. 501968661
Encompass_Preferred
Medchemexpress, HY-109055 10mg Elenbecestat CAS:1388651-30-6 Elenbecestat (E2609) is a potent, orally bioavailable and CNS-penetrant BACE-1 inhibitor for the treatment of Alzheimer's disease (AD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8661 Supplier Medchemexpress LLC Supplier No. HY10905510MG
Medchemexpress, HY-109055 10mg Elenbecestat CAS:1388651-30-6 Elenbecestat (E2609) is a potent, orally bioavailable and CNS-penetrant BACE-1 inhibitor for the treatment of Alzheimer's disease (AD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.