Learn More
Selleck Chemical LLC Elenbecestat
SDP
Catalog No. 5000002856
Encompass_Preferred
Elenbecestat (E2609) is a novel BACE1 inhibitor demonstrating prolonged reductions in plasma beta-amyloid levels after single dosing
Catalog No. 50-000-02856 Supplier Selleck Chemical LLC Supplier No. S86002MG
Elenbecestat (E2609) is a novel BACE1 inhibitor demonstrating prolonged reductions in plasma beta-amyloid levels after single dosing