Learn More
Selleck Chemical LLC Elenestinib phosphate-25MG
SDP
Catalog No. 503136624
Encompass_Preferred
Elenestinib phosphate(BLU-263 phosphate) is a potent and orally active inhibitor of tyrosine kinase that exhibits potent inhibition of c-KIT (D816V) mutation BLU-263 phosphate has the potential for its use in the research of systemic mastocytosis (SM)
Catalog No. 50-313-6624 Supplier Selleck Chemical LLC Supplier No. E166225MG
Elenestinib phosphate(BLU-263 phosphate) is a potent and orally active inhibitor of tyrosine kinase that exhibits potent inhibition of c-KIT (D816V) mutation BLU-263 phosphate has the potential for its use in the research of systemic mastocytosis (SM)